Assay Details
Gene Symbol: Gfra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012959.1 | NP_037091.1 | 7 - 8 | 1270 | 99 | |
XM_008760512.1 | XP_008758734.1 | 8 - 9 | 1413 | 99 | ||
XM_008760513.1 | XP_008758735.1 | 8 - 9 | 1277 | 99 | ||
XM_008760514.1 | XP_008758736.1 | 6 - 7 | 1293 | 99 | ||
GenBank mRNA | AJ002072.1 | - | 6 - 7 | 999 | 99 | |
U59486.1 | - | 7 - 8 | 1315 | 99 | ||
U97142.1 | - | 7 - 8 | 1270 | 99 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
