Assay Details
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031643.4 | NP_113831.1 | 5 - 6 | 622 | 147 | |
GenBank mRNA | BC089772.1 | - | 5 - 6 | 616 | 7382030 | 147 |
D13341.1 | - | 5 - 6 | 605 | 147 | ||
D14591.1 | - | 5 - 6 | 575 | 147 | ||
X62313.1 | - | 5 - 6 | 612 | 147 | ||
Z16415.1 | - | 5 - 6 | 605 | 147 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
