Assay Details
Gene Symbol: Map2k1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031643.4 | NP_113831.1 | 9 - 10 | 1060 | 97 | |
GenBank mRNA | BC089772.1 | - | 9 - 10 | 1054 | 97 | |
D13341.1 | - | 9 - 10 | 1043 | 97 | ||
D14591.1 | - | 9 - 10 | 1013 | 97 | ||
X62313.1 | - | 9 - 10 | 1050 | 97 | ||
Z16415.1 | - | 9 - 10 | 1043 | 97 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
