Assay Details
Gene Symbol: Usp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006242861.2 | XP_006242923.1 | 1 - 2 | 244 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006242861.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
