Assay Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024394.2 | NP_077370.2 | 5 - 6 | 670 | 89 | |
XM_006243021.2 | XP_006243083.1 | 5 - 6 | 684 | 89 | ||
GenBank mRNA | D49395.1 | - | 5 - 6 | 561 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
