Assay Details
Gene Symbol: Htr3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_024394.2 | NP_077370.2 | 7 - 8 | 1040 | 73 | |
XM_006243021.2 | XP_006243083.1 | 7 - 8 | 1054 | 73 | ||
GenBank mRNA | D49395.1 | - | 7 - 8 | 931 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
