Assay Details
Gene Symbol: Cadm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012201.1 | NP_001012201.1 | 4 - 5 | 736 | 79 | |
XM_006242944.2 | XP_006243006.1 | 4 - 5 | 735 | 79 | ||
XM_006242946.2 | XP_006243008.1 | 4 - 5 | 735 | 79 | ||
XM_006242947.2 | XP_006243009.1 | 4 - 5 | 735 | 79 | ||
XM_006242948.2 | XP_006243010.1 | 4 - 5 | 735 | 79 | ||
GenBank mRNA | AB114443.1 | - | 4 - 5 | 625 | 79 | |
BC078966.1 | - | 4 - 5 | 736 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
