Assay Details
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017075.2 | NP_058771.2 | 6 - 7 | 655 | 79 | |
GenBank mRNA | D00512.1 | - | 6 - 7 | 587 | 79 | |
D13921.1 | - | 6 - 7 | 587 | 79 | ||
FM071064.1 | - | 4 - 5 | 443 | 79 | ||
FQ217320.1 | - | 6 - 7 | 593 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
