Assay Details
Gene Symbol: Acat1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017075.2 | NP_058771.2 | 7 - 8 | 809 | 97 | |
GenBank mRNA | D00512.1 | - | 7 - 8 | 741 | 97 | |
D13921.1 | - | 7 - 8 | 741 | 97 | ||
FM098848.1 | - | 2 - 3 | 191 | 97 | ||
FQ217320.1 | - | 7 - 8 | 747 | 97 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
