Assay Details
Gene Symbol: Mapk6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031622.2 | NP_113810.2 | 1 - 2 | 143 | 70 | |
GenBank mRNA | BC109380.1 | - | 1 - 2 | 94 | 70 | |
CB742965.1 | - | 1 - 2 | 203 | 70 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC109380.1, NM_031622.2 transcripts.
This assay is in the 5'UTR region of the BC109380.1, NM_031622.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
