Assay Details
Gene Symbol: Tgfbr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766690.1 | XP_008764912.1 | 6 - 7 | 2203 | 86 | |
GenBank mRNA | L09653.1 | - | 5 - 6 | 1454 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
