Assay Details
Gene Symbol: Slc25a38 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030032.1 | NP_001025203.1 | 4 - 5 | 601 | 63 | |
XM_006244089.2 | XP_006244151.1 | 4 - 5 | 2502 | 63 | ||
GenBank mRNA | BC099762.1 | - | 4 - 5 | 601 | 63 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
