Assay Details
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047916.1 | NP_001041381.1 | 3 - 4 | 257 | 82 | |
XM_006243513.2 | XP_006243575.1 | 995 | 82 | |||
XM_006243514.1 | XP_006243576.1 | 950 | 82 | |||
XM_006243515.1 | XP_006243577.1 | 950 | 82 | |||
XM_006243516.1 | XP_006243578.1 | 950 | 82 | |||
XM_006243517.2 | XP_006243579.1 | 2 - 3 | 222 | 82 | ||
XM_006243518.2 | XP_006243580.1 | 4 - 5 | 446 | 82 | ||
XM_006243519.2 | XP_006243581.1 | 4 - 5 | 446 | 82 | ||
GenBank mRNA | AY325202.1 | - | 3 - 4 | 257 | 82 |
Gene Symbol: LOC100910033 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243521.2 | XP_006243583.1 | 1318 | 82 |
Gene Symbol: LOC100910069 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008766471.1 | XP_008764693.1 | 7 - 8 | 1255 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
