Assay Details
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 3 - 4 | 227 | 116 | |
XM_008766440.1 | XP_008764662.1 | 3 - 4 | 278 | 116 | ||
GenBank mRNA | BC085352.1 | - | 3 - 4 | 242 | 116 | |
FQ210730.1 | - | 3 - 4 | 237 | 116 | ||
FQ213183.1 | - | 3 - 4 | 237 | 116 | ||
FQ219177.1 | - | 3 - 4 | 237 | 116 | ||
FQ219285.1 | - | 3 - 4 | 237 | 116 | ||
FQ219548.1 | - | 3 - 4 | 237 | 116 | ||
FQ219611.1 | - | 3 - 4 | 255 | 116 | ||
FQ223707.1 | - | 3 - 4 | 233 | 116 | ||
FQ228686.1 | - | 3 - 4 | 297 | 116 | ||
FQ228906.1 | - | 3 - 4 | 244 | 116 | ||
FQ228938.1 | - | 3 - 4 | 234 | 116 | ||
FQ228956.1 | - | 3 - 4 | 264 | 116 | ||
M38135.1 | - | 3 - 4 | 221 | 116 | ||
Y00708.1 | - | 3 - 4 | 227 | 116 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
