Assay Details
Gene Symbol: Ctsh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_012939.1 | NP_037071.1 | 5 - 6 | 406 | 83 | |
XM_008766440.1 | XP_008764662.1 | 5 - 6 | 457 | 83 | ||
GenBank mRNA | BC085352.1 | - | 5 - 6 | 421 | 7115929 | 83 |
FQ210730.1 | - | 5 - 6 | 416 | 83 | ||
FQ213183.1 | - | 5 - 6 | 416 | 83 | ||
FQ219177.1 | - | 5 - 6 | 416 | 83 | ||
FQ219281.1 | - | 5 - 6 | 416 | 83 | ||
FQ219285.1 | - | 5 - 6 | 416 | 83 | ||
FQ219548.1 | - | 5 - 6 | 416 | 83 | ||
FQ219611.1 | - | 5 - 6 | 434 | 83 | ||
FQ220588.1 | - | 2 - 3 | 185 | 83 | ||
FQ228686.1 | - | 5 - 6 | 476 | 83 | ||
FQ228938.1 | - | 5 - 6 | 413 | 83 | ||
FQ228956.1 | - | 5 - 6 | 443 | 83 | ||
M38135.1 | - | 5 - 6 | 400 | 83 | ||
Y00708.1 | - | 5 - 6 | 406 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
