Assay Details
Gene Symbol: Hemk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106853.1 | NP_001100323.1 | 9 - 10 | 1060 | 58 | |
XM_006243728.2 | XP_006243790.1 | 10 - 11 | 1350 | 58 | ||
XM_006243729.2 | XP_006243791.1 | 10 - 11 | 1670 | 58 | ||
XM_006243730.2 | XP_006243792.1 | 9 - 10 | 1425 | 58 | ||
XM_008766502.1 | XP_008764724.1 | 10 - 11 | 1285 | 58 | ||
GenBank mRNA | BC161944.1 | - | 9 - 10 | 1054 | 58 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
