Assay Details
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133427.3 | NP_596918.3 | 10 - 11 | 948 | 83 | |
XM_006243472.2 | XP_006243534.1 | 9 - 10 | 934 | 83 | ||
XM_008766437.1 | XP_008764659.1 | 10 - 11 | 1107 | 83 | ||
GenBank mRNA | AF307840.1 | - | 9 - 10 | 707 | 83 | |
AY321370.1 | - | 9 - 10 | 707 | 83 | ||
AY321371.1 | - | 9 - 10 | 707 | 83 | ||
BC080240.1 | - | 10 - 11 | 926 | 7099950 | 83 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
