Assay Details
Gene Symbol: Cyb5r4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133427.3 | NP_596918.3 | 1 - 2 | 213 | 100 | |
XM_006243472.2 | XP_006243534.1 | 1 - 2 | 229 | 100 | ||
GenBank mRNA | BC080240.1 | - | 1 - 2 | 191 | 7099950 | 100 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC080240.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
