Assay Details
Gene Symbol: Eef1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175838.1 | NP_787032.1 | 3 - 4 | 396 | 82 | |
GenBank mRNA | BC063162.1 | - | 3 - 4 | 392 | 82 | |
BC072542.1 | - | 3 - 4 | 391 | 82 | ||
BC091297.1 | - | 3 - 4 | 377 | 82 | ||
BC111707.1 | - | 2 - 3 | 360 | 82 | ||
BC128723.1 | - | 3 - 4 | 371 | 82 | ||
L10339.1 | - | 2 - 3 | 331 | 82 | ||
X61043.1 | - | 3 - 4 | 396 | 82 | ||
X63561.1 | - | 2 - 3 | 369 | 82 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
