Assay Details
Gene Symbol: Mobp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF157498.1 | - | 6 - 7 | 912 | 74 | |
BC087694.1 | - | 4 - 5 | 570 | 74 | ||
D28111.1 | - | 5 - 6 | 696 | 74 | ||
X87900.1 | - | 5 - 6 | 609 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
