Assay Details
Gene Symbol: Tmem30a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001004248.1 | NP_001004248.1 | 4 - 5 | 878 | 142 | |
XM_006243435.2 | XP_006243497.1 | 5 - 6 | 988 | 142 | ||
GenBank mRNA | BC079203.1 | - | 4 - 5 | 878 | 142 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
