Assay Details
Gene Symbol: Leo1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005548.1 | NP_001005548.1 | 10 - 11 | 1844 | 96 | |
GenBank mRNA | BC082098.1 | - | 10 - 11 | 1844 | 96 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
