Assay Details
Gene Symbol: Crtap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108785.1 | NP_001102255.1 | 1 - 2 | 551 | 91 | |
XM_006243992.2 | XP_006244054.1 | 1 - 2 | 564 | 91 |
Gene Symbol: LOC102555086 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006244022.2 | XP_006244084.1 | 562 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
