Assay Details
Gene Symbol: P4htm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_217285.8 | XP_217285.6 | 5 - 6 | 1554 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
