Assay Details
Gene Symbol: Stac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_343491.7 | XP_343492.4 | 1 - 2 | 353 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
