Assay Details
Gene Symbol: Stac | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_343491.7 | XP_343492.4 | 4 - 5 | 812 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
