Assay Details
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106852.1 | NP_001100322.1 | 6 - 7 | 807 | 63 | |
GenBank mRNA | FQ233139.1 | - | 2 - 3 | 161 | 63 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
