Assay Details
Gene Symbol: Mrpl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106852.1 | NP_001100322.1 | 8 - 9 | 999 | 87 | |
GenBank mRNA | FQ233139.1 | - | 4 - 5 | 353 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
