Assay Details
Gene Symbol: Nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191884.1 | NP_001178813.1 | 2 - 3 | 217 | 68 | |
XM_006243847.2 | XP_006243909.1 | 2 - 3 | 226 | 68 | ||
XM_006243848.2 | XP_006243910.1 | 2 - 3 | 152 | 68 | ||
XM_006243849.1 | XP_006243911.1 | 2 - 3 | 670 | 68 | ||
XM_006243850.2 | XP_006243912.1 | 2 - 3 | 191 | 68 | ||
XM_006243851.1 | XP_006243913.1 | 2 - 3 | 255 | 68 | ||
GenBank mRNA | FQ225386.1 | - | 1 - 2 | 147 | 68 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001191884.1, XM_006243847.2, XM_006243848.2, XM_006243849.1, XM_006243850.2, XM_006243851.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
