Assay Details
Gene Symbol: Nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191884.1 | NP_001178813.1 | 5 - 6 | 465 | 66 | |
XM_006243847.2 | XP_006243909.1 | 5 - 6 | 474 | 66 | ||
XM_006243848.2 | XP_006243910.1 | 5 - 6 | 400 | 66 | ||
XM_006243849.1 | XP_006243911.1 | 5 - 6 | 918 | 66 | ||
XM_006243850.2 | XP_006243912.1 | 5 - 6 | 439 | 66 | ||
XM_006243851.1 | XP_006243913.1 | 5 - 6 | 503 | 66 | ||
GenBank mRNA | AF051943.1 | - | 3 - 4 | 207 | 66 | |
FQ225386.1 | - | 5 - 6 | 644 | 66 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
