Assay Details
Gene Symbol: Nme6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191884.1 | NP_001178813.1 | 6 - 7 | 618 | 57 | |
XM_006243847.2 | XP_006243909.1 | 6 - 7 | 627 | 57 | ||
XM_006243848.2 | XP_006243910.1 | 6 - 7 | 553 | 57 | ||
XM_006243849.1 | XP_006243911.1 | 6 - 7 | 1071 | 57 | ||
XM_006243850.2 | XP_006243912.1 | 6 - 7 | 592 | 57 | ||
XM_006243851.1 | XP_006243913.1 | 6 - 7 | 656 | 57 | ||
GenBank mRNA | AF051943.1 | - | 4 - 5 | 360 | 57 | |
FQ225386.1 | - | 6 - 7 | 797 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
