Assay Details
Gene Symbol: Atrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106859.1 | NP_001100329.1 | 5 - 6 | 936 | 68 | |
XM_006243764.2 | XP_006243826.1 | 5 - 6 | 938 | 68 | ||
XM_006243766.2 | XP_006243828.1 | 5 - 6 | 937 | 68 | ||
XM_006243767.2 | XP_006243829.1 | 5 - 6 | 937 | 68 |
Gene Symbol: LOC100911807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750589.3 | XP_003750637.1 | 5 - 6 | 1931 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
