Assay Details
Gene Symbol: Atrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106859.1 | NP_001100329.1 | 6 - 7 | 1030 | 79 | |
XM_006243764.2 | XP_006243826.1 | 6 - 7 | 1032 | 79 | ||
XM_006243766.2 | XP_006243828.1 | 6 - 7 | 1031 | 79 | ||
XM_006243767.2 | XP_006243829.1 | 6 - 7 | 1031 | 79 |
Gene Symbol: LOC100911807 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003750589.3 | XP_003750637.1 | 6 - 7 | 2025 | 79 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
