Assay Details
Gene Symbol: Acy1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005383.1 | NP_001005383.1 | 11 - 12 | 916 | 77 | |
XM_006243715.2 | XP_006243777.1 | 12 - 13 | 1218 | 77 | ||
XM_006243716.2 | XP_006243778.1 | 12 - 13 | 1222 | 77 | ||
GenBank mRNA | AY580164.1 | - | 11 - 12 | 916 | 77 | |
AY580165.1 | - | 11 - 12 | 988 | 77 | ||
BC078930.1 | - | 12 - 13 | 979 | 77 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
