Assay Details
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 14 - 15 | 2196 | 87 | |
XM_006243872.2 | XP_006243934.1 | 14 - 15 | 2195 | 87 | ||
XM_006243880.2 | XP_006243942.1 | 10 - 11 | 1807 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
