Assay Details
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 16 - 17 | 2403 | 59 | |
XM_006243872.2 | XP_006243934.1 | 16 - 17 | 2402 | 59 | ||
XM_006243880.2 | XP_006243942.1 | 12 - 13 | 2014 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
