Assay Details
Gene Symbol: Cdhr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243871.2 | XP_006243933.1 | 1 - 2 | 255 | 92 | |
XM_006243872.2 | XP_006243934.1 | 1 - 2 | 254 | 92 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
