Assay Details
Gene Symbol: Hmgcll1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276472.1 | NP_001263401.1 | 6 - 7 | 718 | 87 | |
XM_006243415.2 | XP_006243477.1 | 6 - 7 | 1387 | 87 | ||
GenBank mRNA | FQ211656.1 | - | 6 - 7 | 718 | 87 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
