Assay Details
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019267.1 | NP_062140.1 | 2 - 3 | 296 | 112 | |
XM_006243450.2 | XP_006243512.1 | 3 - 4 | 350 | 112 | ||
GenBank mRNA | BC158861.1 | - | 2 - 3 | 289 | 7932093 | 112 |
CK473898.1 | - | 2 - 3 | 296 | 112 | ||
FQ213896.1 | - | 2 - 3 | 278 | 112 | ||
FQ226240.1 | - | 2 - 3 | 279 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
