Assay Details
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019267.1 | NP_062140.1 | 3 - 4 | 367 | 92 | |
XM_006243450.2 | XP_006243512.1 | 4 - 5 | 421 | 92 | ||
GenBank mRNA | BC158861.1 | - | 3 - 4 | 360 | 92 | |
CK473898.1 | - | 3 - 4 | 367 | 92 | ||
FQ213896.1 | - | 3 - 4 | 349 | 92 | ||
FQ226240.1 | - | 3 - 4 | 350 | 92 | ||
M94040.1 | - | 3 - 4 | 277 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
