Assay Details
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019267.1 | NP_062140.1 | 5 - 6 | 651 | 136 | |
XM_006243450.2 | XP_006243512.1 | 6 - 7 | 708 | 136 | ||
GenBank mRNA | BC158861.1 | - | 5 - 6 | 644 | 7932093 | 136 |
CK473898.1 | - | 5 - 6 | 651 | 136 | ||
FQ213896.1 | - | 5 - 6 | 633 | 136 | ||
M94040.1 | - | 5 - 6 | 562 | 136 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
