Assay Details
Gene Symbol: Bckdhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_019267.1 | NP_062140.1 | 9 - 10 | 1061 | 151 | |
XM_006243450.2 | XP_006243512.1 | 10 - 11 | 1118 | 151 | ||
GenBank mRNA | BC158861.1 | - | 9 - 10 | 1054 | 151 | |
BM385109.1 | - | 2 - 3 | 151 | |||
DV215445.1 | - | 1 - 2 | 80 | 151 | ||
FQ213896.1 | - | 9 - 10 | 1043 | 151 | ||
M94040.1 | - | 9 - 10 | 972 | 151 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
