Assay Details
Gene Symbol: Abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212524.1 | NP_997689.1 | 1 - 2 | 94 | 106 | |
GenBank mRNA | AY550934.1 | - | 1 - 2 | 94 | 106 | |
BC127470.1 | - | 1 - 2 | 128 | 106 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AY550934.1,NM_212524.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
