Assay Details
Gene Symbol: Abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212524.1 | NP_997689.1 | 2 - 3 | 180 | 89 | |
XM_006244107.1 | XP_006244169.1 | 3 - 4 | 285 | 89 | ||
XM_006244108.2 | XP_006244170.1 | 2 - 3 | 185 | 89 | ||
GenBank mRNA | AY550934.1 | - | 2 - 3 | 180 | 89 | |
BC127470.1 | - | 2 - 3 | 214 | 89 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006244107.1, XM_006244108.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
