Assay Details
Gene Symbol: Abhd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212524.1 | NP_997689.1 | 3 - 4 | 549 | 75 | |
XM_006244107.1 | XP_006244169.1 | 4 - 5 | 654 | 75 | ||
XM_006244108.2 | XP_006244170.1 | 3 - 4 | 554 | 75 | ||
XM_006244109.2 | XP_006244171.1 | 2 - 3 | 1942 | 75 | ||
GenBank mRNA | AY550934.1 | - | 3 - 4 | 549 | 75 | |
BC127470.1 | - | 3 - 4 | 583 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
