Assay Details
Gene Symbol: Nck1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106851.2 | NP_001100321.1 | 1 - 2 | 122 | 120 | |
GenBank mRNA | BC167009.1 | - | 1 - 2 | 122 | 120 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC167009.1, NM_001106851.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
