Assay Details
Gene Symbol: Gmppb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108781.1 | NP_001102251.1 | 7 - 8 | 997 | 65 | |
XM_006243811.2 | XP_006243873.1 | 8 - 9 | 1294 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
