Assay Details
Gene Symbol: Srprb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013252.1 | NP_001013270.1 | 5 - 6 | 568 | 70 | |
GenBank mRNA | AY325214.1 | - | 2716 | 70 | ||
AY325230.1 | - | 2719 | 70 | |||
AY325241.1 | - | 2716 | 70 | |||
BC098951.1 | - | 560 | 70 | |||
BF523222.1 | - | 2 - 3 | 234 | 70 | ||
BQ204036.1 | - | 2 - 3 | 70 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
