Assay Details
Gene Symbol: Filip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145682.1 | NP_663715.1 | 1 - 2 | 69 | 73 | |
GenBank mRNA | AB055759.1 | - | 1 - 2 | 69 | 73 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the AB055759.1, NM_145682.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
