Assay Details
Gene Symbol: Tex264 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007665.1 | NP_001007666.1 | 2 - 3 | 216 | 91 | |
XM_006243726.2 | XP_006243788.1 | 2 - 3 | 182 | 91 | ||
XM_006243727.2 | XP_006243789.1 | 2 - 3 | 483 | 91 | ||
XM_008766501.1 | XP_008764723.1 | 2 - 3 | 172 | 91 | ||
GenBank mRNA | BC083732.1 | - | 2 - 3 | 216 | 7191345 | 91 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC083732.1, NM_001007665.1, XM_006243726.2, XM_006243727.2, XM_008766501.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
MicroArray Probes:
